Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCL6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541693
Description
Peptide sequence: LQSDLDQPAF QQLVSFSESG SLGNSSGSDV TSLSSQLPDT PNSMVPSPVE Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
BCL6B acts as a sequence-specific transcriptional repressor in association with BCL6. BCL6B may function in a narrow stage or be related to some events in the early B-cell development.
Specifications
| BCL6B | |
| Polyclonal | |
| Unconjugated | |
| BCL6B | |
| B cell CLL/lymphoma 6, member B; B cell CLL/lymphoma 6B; BAZF; B-cell CLL/lymphoma 6 member B protein; B-cell CLL/lymphoma 6, member B; B-cell CLL/lymphoma 6, member B (zinc finger protein); B-cell CLL/lymphoma 6B; Bcl6-associated zinc finger protein; Bcl6b; BCL6B transcription repressor; BCL6B, transcription repressor; LOW QUALITY PROTEIN: B-cell CLL/lymphoma 6 member B protein; RGD1563179; ZBTB28; zinc finger protein 62; ZNF62 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 255877 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N143 | |
| BCL6B | |
| Synthetic peptide directed towards the middle region of human BCL6B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction