Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983017
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3017 Supplier Bioss Supplier No. BS13606R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL6B acts as a sequence-specific transcriptional repressor in association with BCL6. BCL6B may function in a narrow stage or be related to some events in the early B-cell development.

Specifications

Antigen BCL6B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BCL6B
Gene Accession No. Q8N143
Gene Alias B cell CLL/lymphoma 6, member B; B cell CLL/lymphoma 6B; BAZF; B-cell CLL/lymphoma 6 member B protein; B-cell CLL/lymphoma 6, member B; B-cell CLL/lymphoma 6, member B (zinc finger protein); B-cell CLL/lymphoma 6B; Bcl6-associated zinc finger protein; Bcl6b; BCL6B, transcription repressor; BOS_18750; RGD1563179; ZBTB28; Zinc finger protein 62; ZNF62
Gene Symbols BCL6B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BCL6B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255877
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.