Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL7B Antibody (OTI7H1), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP344785
Description
BCL7B Monoclonal antibody specifically detects BCL7B in Human,Mouse samples. It is validated for Western BlotSpecifications
BCL7B | |
Monoclonal | |
Unconjugated | |
Lyophilized powder ( 1X PBS, pH 7.3,) | |
Mouse | |
>95% | |
RUO | |
9275 | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
OTI7H1 | |
Western Blot 1:2000 | |
Allergen Hom s 3, B-cell CLL/lymphoma 7 protein family member B, B-cell CLL/lymphoma 7B | |
Full length human recombinant protein of human BCL7B (NP_001698) produced in E.coli. | |
100 μg | |
Primary | |
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction