Learn More
BCL7B Mouse anti-Human, Clone: OTI2D9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809485

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.Specifications
BCL7B | |
Monoclonal | |
Unconjugated | |
BCL7B | |
BCL tumor suppressor 7B | |
Mouse | |
Affinity Chromatography | |
RUO | |
9275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BQE9 | |
BCL7B | |
Full length human recombit protein of human BCL7B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.