Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL7C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553841 Supplier Invitrogen™ Supplier No. PA553841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMLTKEEPVP ELLEAEDSGV RMTRRALHEK GLKTEPLRRL LPRRGLRTNV RPSSMAVPDT RAPGGGSKAP RAPRTT Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 31%.

This gene is identified by the similarity of its product to the N-terminal region of BCL7A protein. The BCL7A protein is encoded by the gene known to be directly involved in a three-way gene translocation in a Burkitt lymphoma cell line. The function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL7C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCL7C
Gene Accession No. Q8WUZ0
Gene Alias B cell CLL/lymphoma 7C; B-cell chronic lymphocytic leukemia/lymphoma 7C protein; B-cell CLL/lymphoma 7 protein family member C; B-cell CLL/lymphoma 7C; BCL tumor suppressor 7C; Bcl7c; C230096E12Rik
Gene Symbols BCL7C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCL7C. Recombinant protein control fragment (Product #RP-92093).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.