Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL7C Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7249083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170444 25 μg
NB170437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB170444 Supplier Novus Biologicals Supplier No. NBP31864225UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL7C Polyclonal antibody specifically detects BCL7C in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen BCL7C
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:1000
Formulation Whole antiserum
Gene Accession No. Q8WUZ0
Gene Alias B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human BCL7C (B-cell CLL/lymphoma 7C), using two KLH-conjugated synthetic peptides containing a sequence from the central and from the C-terminal part of the protein, respectively.
Purification Method Whole Antiserum
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9274
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.