Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL9-2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171907AF750
Description
BCL9-2 Polyclonal antibody specifically detects BCL9-2 in Human samples. It is validated for Western BlotSpecifications
BCL9-2 | |
Polyclonal | |
Western Blot | |
B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, BCL9-like protein, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2 | |
The immunogen for this product maps to a region between residue 525 and 575 of human B-Cell CLL/Lymphoma 9-Like using the numbering given in entry NP_872363.1 (GeneID 283149). | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
283149 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction