Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549466
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549466 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549466 Supplier Invitrogen™ Supplier No. PA549466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL9 Polyclonal Antibody for Western Blot, ICC/IF

BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin's transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BCL9
Gene Accession No. O00512, Q9D219
Gene Alias 2610202E01Rik; 8030475K17Rik; A330041G23Rik; B cell CLL/lymphoma 9; B-cell CLL/lymphoma 9; B-cell CLL/lymphoma 9 protein; B-cell lymphoma 9 protein; BCL9; bcl-9; Gm130; LGS; nuclear co-factor of beta-catenin signalling; protein legless homolog
Gene Symbols BCL9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 272-304 amino acids from human BCL9.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 607, 77578
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.