Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597503
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597503 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597503 Supplier Invitrogen™ Supplier No. PA597503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin's transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BCL9
Gene Accession No. O00512
Gene Alias 2610202E01Rik; 8030475K17Rik; A330041G23Rik; B cell CLL/lymphoma 9; B-cell CLL/lymphoma 9; B-cell CLL/lymphoma 9 protein; B-cell lymphoma 9 protein; BCL9; bcl-9; Gm130; LGS; nuclear co-factor of beta-catenin signalling; protein legless homolog
Gene Symbols BCL9
Host Species Rabbit
Immunogen Recombinant Human B-cell CLL/lymphoma 9 protein (231-340AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 607
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.