Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA576568
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin's transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell malignancies.
Specifications
BCL9 | |
Polyclonal | |
Unconjugated | |
BCL9 | |
2610202E01Rik; 8030475K17Rik; A330041G23Rik; B cell CLL/lymphoma 9; B-cell CLL/lymphoma 9; B-cell CLL/lymphoma 9 protein; B-cell lymphoma 9 protein; BCL9; bcl-9; Gm130; LGS; nuclear co-factor of beta-catenin signalling; protein legless homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
310704, 607, 77578 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O00512, Q9D219 | |
BCL9 | |
Recombinant protein of human Bcl9 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction