Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL9 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733971 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3971 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3971 Supplier Proteintech Group Inc Supplier No. 229471AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin′s transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell maligcies

Specifications

Antigen BCL9
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCL9
Gene Accession No. O00512
Gene Alias B cell CLL/lymphoma 9, B cell CLL/lymphoma 9 protein, B cell lymphoma 9 protein, BCL9, LGS, Protein legless homolog
Gene Symbols BCL9
Host Species Rabbit
Immunogen BCL9 Fusion Protein Ag19158
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 607
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.