Learn More
Invitrogen™ BCL9L Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-21111 can be used with blocking peptide PEP-1225.
Specifications
Specifications
| Antigen | BCL9L |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BCL9L |
| Gene Accession No. | Q86UU0 |
| Gene Alias | B cell CLL/lymphoma 9-like; B9L; BC003321; B-cell CLL/lymphoma 9-like; B-cell CLL/lymphoma 9-like protein; B-cell lymphoma 9-like protein; BCL9-2; BCL9L; BCL9-like protein; BCL9-related beta-catenin-binding protein; DLNB11; nuclear co-factor of beta-catenin signalling; protein BCL9-2 |
| Gene Symbols | BCL9L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.