Learn More
BCLAF1 Mouse, anti-Human, Unconjugated, Clone: 1D6G7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | BCLAF1 |
---|---|
Clone | 1D6G7 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9902
![]() ![]() |
Proteintech Group Inc
678601IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
BCLF1, also named as Bcl-2-associated transcription factor 1, is a 920 amino acid protein, which Interacts with Bcl-2 related proteins, EMD, with the adenovirus E1B 19 kDa protein and with DNA. BCLF1 as a death-promoting transcriptional repressor may be involved in cyclin-D1/CCND1 mRNA stability through the SNARP complex which associates with both the 3′end of the CCND1 gene and its mRNA. The calculated molecular weight of BCLF1 is a 110 kDa, but the modified BCLF1 protein is about 140-150 kDa.
Specifications
BCLAF1 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human | |
Primary |
1D6G7 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse | |
9774 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.