Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMA/TNFRSF17 Antibody (049), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312791MFV500
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISASpecifications
| BCMA/TNFRSF17 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 049 | |
| ELISA | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BCMA/TNFRSF17 (Accession#: NP_001183.2; Met1-Ala54). | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction | |
| 608 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction