Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMA/TNFRSF17 Antibody (Vicky-1) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
$269.00 - $426.00
Specifications
Antigen | BCMA/TNFRSF17 |
---|---|
Clone | Vicky-1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Blocking/Neutralizing 50-200 molar excess |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Blocking Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197637SS
![]() |
Novus Biologicals
NBP197637SS |
0.025 mg |
Each for $269.00
|
|
|||||
NBP197637
![]() |
Novus Biologicals
NBP197637 |
0.05 mg |
Each for $426.00
|
|
|||||
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
BCMA/TNFRSF17 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Blocking Assay | |
Unconjugated | |
Rat | |
Human | |
608 | |
Recombinant human BCMA extracellular domain (aa 2-54). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Vicky-1 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Blocking/Neutralizing 50-200 molar excess | |
Monoclonal | |
Purified | |
RUO | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
TNFRSF17 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title