Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMA/TNFRSF17 Antibody (Vicky-1), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP197637UV0.1ML
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
BCMA/TNFRSF17 | |
Monoclonal | |
DyLight 350 | |
50 mM Sodium Borate | |
TNFRSF17 | |
Recombinant human BCMA extracellular domain (aa 2-54). | |
Cytokine Research, Immunology | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Vicky-1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
Rat | |
0.1 mL | |
608 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction