Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMA/TNFRSF17 Antibody (Vicky-1), FITC, Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP197655100TESTS
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.Specifications
BCMA/TNFRSF17 | |
Monoclonal | |
FITC | |
PBS | |
TNFRSF17 | |
Recombinant human BCMA extracellular domain (aa 2-54). | |
608 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry | |
Vicky-1 | |
Flow Cytometry 1:20 | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
Rat | |
100 Tests | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction