Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590759 Supplier Invitrogen™ Supplier No. PA590759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSWASCLAFH REEKTYIHII DQRTRQPVQT KFYTDAMVVF HHVNAYEEDG CIVFDVIAYE DNSLYQLFYL ANLNQDFKEN SRLTSVPTLR RFAVPLHVDK NAEVGTNLIK VASTTATALK EEDGQVYCQP EFLYEGLELP RVNYAHNGKQ YRYVFATGVQ WSPIPTKIIK YDILTKSSLK WREDDCWPAE PLFVPAPGAK DEDDGVILSA IVSTDPQKLP FLLILDAKSF TELARASVDV DMHMDLHGLF ITDMDWDTKK QAASEEQRDR ASDCHGAPLT; Positive Samples: HT-29, 293T, LO2, Mouse liver, Mouse testis.

Vitamin A metabolism is important for vital processes such as vision, embryonic development, cell differentiation, and membrane and skin protection. The protein encoded by this gene is a key enzyme in beta-carotene metabolism to vitamin A. It catalyzes the oxidative cleavage of beta, beta-carotene into two retinal molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCMO1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BCO1
Gene Accession No. Q91XT5, Q9HAY6, Q9JJS6
Gene Alias BCDO; BCDO1; BCMO; Bcmo1; BCO; BCO1; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15 15'-dioxygenase; beta-carotene 15 15-dioxygenase; beta-carotene 15, 15'-dioxygenase; beta-carotene 15, 15-dioxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15' dioxygenase; beta-carotene 15,15'-monooxygenase; beta-carotene 15,15'-monooxygenase 1; beta-carotene cleavage enzyme; beta-carotene dioxygenase 1; Beta-carotene oxygenase 1; beta-CD; betaCMOOX; carotenoid-15,15'-oxygenase; CMO1; Cmoi
Gene Symbols BCO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 268-547 of human BCMO1 (NP_0591252).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114106, 53630, 63857
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.