Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMP101 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8211R

Description
FAM84B is a protein coding gene.Specifications
BCMP101 | |
Polyclonal | |
Unconjugated | |
FAM84B | |
BCMP101; Breast cancer membrane protein 101; breast cancer membrane-associated protein 101; FAM84B; family with sequence similarity 84 member B; family with sequence similarity 84, member B; LRAT domain-containing 2; LRATD2; neurological/sensory 2; NSE2; Protein FAM84B; Protein LRATD2; Protein NSE2 | |
Rabbit | |
Protein A | |
RUO | |
157638 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96KN1 | |
FAM84B | |
KLH conjugated synthetic peptide derived from human FAM84B/BCMP101. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction