Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140101 Supplier Invitrogen™ Supplier No. PA5140101
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AKSFPRRFVL PLNVSLNAPE GDNLSPLSYT SASAVKQADG TIWCSHENLH QEDLEKEGGI EFPQIYYDRF SGKKYHFFYG CG Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 79%.

BCDO2 catalyzes the asymmetric oxidative cleavage of beta-carotene in carotene metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCO2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCO2
Gene Accession No. Q9BYV7
Gene Alias Ab2-079; b,b-carotene-9',10'-dioxygenase; BCDO2; Bcmo2; Bco2; B-diox-II; beta,beta-carotene 9',10'-dioxygenase variant 1; beta,beta-carotene 9',10'-dioxygenase variant 2; beta,beta-carotene 9',10'-dioxygenase variant 3; beta,beta-carotene 9',10'-oxygenase; beta-carotene 9', 10'-dioxygenase 2; beta-carotene 9',10' oxygenase; beta-carotene 9',10'-dioxygenase 2; beta-carotene dioxygenase 2; beta-carotene oxygenase 2; beta-diox-II; carotenoid 9',10' monooxygenase II; carotenoid 9',10' monoxygenase II; carotenoid-9',10'-cleaving dioxygenase; CMO2
Gene Symbols BCO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCO2. Recombinant protein control fragment (Product #RP-109002).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.