Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCOR Antibody (CL13841) [Alexa Fluor 700], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP343828AF700
Description
BCOR Monoclonal antibody specifically detects BCOR in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
BCOR | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
54880 | |
Store at 4°C in the dark. | |
IgG2b |
Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
CL13841 | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
ANOP2, BCL6 corepressor, BCL-6 corepressor, BCL6 co-repressor, BCL-6 interacting corepressor, BCoR, FLJ20285, FLJ38041, KIAA1575MGC71031, MAA2, MCOPS2, MGC131961 | |
This antibody was generated using a synthetic peptide of Q6W2J9, with the exact immunogen sequence remaining proprietary. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction