Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCOR Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298770H
Description
BCOR Polyclonal antibody specifically detects BCOR in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
BCOR | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
ANOP2, BCL6 corepressor, BCL-6 corepressor, BCL6 co-repressor, BCL-6 interacting corepressor, BCoR, FLJ20285, FLJ38041, KIAA1575MGC71031, MAA2, MCOPS2, MGC131961 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human BCOR. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54880 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction