Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCOR Antibody (OTI4F6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271973H
Description
BCOR Monoclonal specifically detects BCOR in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BCOR | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BCOR | |
Recombinant protein fragment corresponding to amino acids 384-668 of human BCOR (NP_060215) produced in E.coli. | |
0.1 mL | |
54880.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4F6 | |
Western Blot | |
ANOP2, BCL6 corepressor, BCL-6 corepressor, BCL6 co-repressor, BCL-6 interacting corepressor, BCoR, FLJ20285, FLJ38041, KIAA1575MGC71031, MAA2, MCOPS2, MGC131961 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction