Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCOR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142066
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142066 Supplier Invitrogen™ Supplier No. PA5142066
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise all reported isoforms (NP_060215.4; NP_001116855.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Transcriptional corepressor. May specifically inhibit gene expression when recruited to promoter regions by sequence-specific DNA-binding proteins such as BCL6 and MLLT3. This repression may be mediated at least in part by histone deacetylase activities which can associate with this corepressor. Involved in the repression of TFAP2A; impairs binding of BCL6 and KDM2B to TFAP2A promoter regions. Via repression of TFAP2A acts as a negative regulator of osteo-dentiogenic capacity in adult stem cells; the function implies inhibition of methylation on histone H3 'Lys-4' (H3K4me3) and 'Lys-36' (H3K36me2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCOR
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Bcor
Gene Accession No. Q6W2J9
Gene Alias 5830466J11Rik; 8430401K06Rik; ANOP2; BCL6 corepressor; BCL-6 corepressor; BCL6 co-repressor; BCL6 interacting corepressor; BCL-6 interacting corepressor; Bcor; BcorR; D930024N20Rik; Kiaa1575; MAA2; MCOPS2; mKIAA1575; RGD1562735
Gene Symbols Bcor
Host Species Goat
Immunogen Peptide with sequence LSATPLYGNVHSW-C (aa 2-14).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54880
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.