Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCORL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556855 Supplier Invitrogen™ Supplier No. PA556855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSSDRIRMCG INEERRAPLS DEESTTGDCQ HFGSQEFCVS SSFSKVELTA VGSGSNARGA DPDGSATEKL GHKSEDKPDD PQPK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.

The protein encoded by this gene is a transcriptional corepressor that is found tethered to promoter regions by DNA-binding proteins. The encoded protein can interact with several different class II histone deacetylases to repress transcription. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCORL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCORL1
Gene Accession No. Q5H9F3
Gene Alias BCL6 corepressor-like 1; BCL-6 corepressor-like protein 1; BCORL1; BCoR-L1; BCoR-like protein 1; CXorf10
Gene Symbols BCORL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCORL1. Recombinant protein control fragment (Product #RP-95578).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.