Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCR/ABL Antibody (4C12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002665AF750
Description
BCR/ABL Monoclonal specifically detects BCR/ABL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
BCR/ABL | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
4C12 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ABL, ALL, BCR ABL fusion protein, bcr/abl, BCR/ABL fusion protein, Bcr/c abl oncogene protein, BCR-ABL fusion protein, BCRe18/ABL1e3 fusion protein, breakpoint cluster region protein, c-ABL, CML, D22S662, JTK7, p150, PHL | |
BCR/ABL peptide | |
0.1 mL | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction