Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCR/ABL Antibody (4C12), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002665C
Description
BCR/ABL Monoclonal specifically detects BCR/ABL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BCR | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BCR | |
BCR/ABL peptide | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
613 | |
Store at 4C in the dark. | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
4C12 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction