Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCR Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119097AF350
Description
BCR Polyclonal antibody specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
BCR | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human breakpoint cluster region using the numbering given in entry NP_004318.3 (GeneID 613). | |
0.1 mL | |
Phospho Specific, Protein Kinase, Signal Transduction | |
613 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunofluorescence | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction