Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100058 Supplier Invitrogen™ Supplier No. PA5100058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BCS1L.

This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Five alternatively spliced transcripts encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCS1L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Bcs1l
Gene Accession No. Q9CZP5, Q9Y276
Gene Alias 9130022O19Rik; BC1 (ubiquinol-cytochrome c reductase) synthesis-like; BCS; BCS1; BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone; Bcs1l; BCS1-like (yeast); BCS1-like protein; BJS; FLNMS; GRACILE; h-BCS; h-BCS1; Hs.6719; MC3DN1; mitochondrial chaperone BCS1; mitochondrial complex III assembly; PTD
Gene Symbols Bcs1l
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCS1L(Accession Q9Y276), corresponding to amino acid residues C234-V284.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301514, 617, 66821
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.