Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD 1008 dihydrobromide, Tocris Bioscience™

Catalog No. 051150
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective σ ligand

Potent and selective σ-ligand (Ki against [3H]-(+)-3-PPP = 0.34 nM).

Chemical Identifiers

CAS 138356-09-9
Molecular Formula C15H24Br2Cl2N2
Molecular Weight (g/mol) 463.079
InChI Key YTBUMHOSKIAYCZ-UHFFFAOYSA-N
Synonym bd 1008 dihydrobromide, n-2-3,4-dichlorophenyl ethyl-n-methyl-2-1-pyrrolidinyl ethylamine dihydrobromide
PubChem CID 45073417
IUPAC Name 2-(3,4-dichlorophenyl)-N-methyl-N-(2-pyrrolidin-1-ylethyl)ethanamine;dihydrobromide
SMILES CN(CCC1=CC(=C(C=C1)Cl)Cl)CCN2CCCC2.Br.Br

Specifications

Quantity 50 mg
Formula Weight 463.08
Percent Purity >98%
Chemical Name or Material BD 1008 dihydrobromide
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.