Learn More
BD Affirm™ VPIII Microbial Identification Test
Click to view available options
Type:
MicroProbe™ Processor
Sample Collection Set
Sample Collection Swab
Test Kit
Description
Includes
Probe Analysis Cards (PAC) individually wrapped containing five beads (negative control, Trichomonas, Gardnerella, Candida, and positive control); and Reagent Cassettes (RC) containing patient sample reservoir, hybridization solution, wash solution, conjugate, and substrate buffer
- Combine with BD Affirm VPIII Ambient Temperature Transport System (Cat. No. 23-025-604)
- For detection and identification of Candida species, Gardnerella vaginalis, and Trichomonas vaginalis nucleic acid in vaginal fluid specimens from vaginitis/vaginosis patients
- Uses two distinct single-stranded nucleic acid probes for each organism: a capture probe and color development probe complementary to unique genetic sequences of target organisms
- Automated processing, with the BD MicroProbe™ Processor (Cat. No. 23-025-600)
- Positive results indicated by visible blue color on target organism bead
- Built-in positive and negative controls on each results card
Specifications
Specifications
Type | Sample Collection Swab |
For Use With (Application) | Detection of vaginitis |
Quantity | 100 Pack |