Learn More
BD Affirm™ VPIII Microbial Identification Test
Catalog No. 23025600
Click to view available options
Type:
MicroProbe™ Processor
Sample Collection Set
Sample Collection Swab
Test Kit
Includes
Probe Analysis Cards (PAC) individually wrapped containing five beads (negative control, Trichomonas, Gardnerella, Candida, and positive control); and Reagent Cassettes (RC) containing patient sample reservoir, hybridization solution, wash solution, conjugate, and substrate buffer
DNA probe molecular test for the detection of vaginitis
  • Combine with BD Affirm VPIII Ambient Temperature Transport System (Cat. No. 23-025-604)
  • For detection and identification of Candida species, Gardnerella vaginalis, and Trichomonas vaginalis nucleic acid in vaginal fluid specimens from vaginitis/vaginosis patients
  • Uses two distinct single-stranded nucleic acid probes for each organism: a capture probe and color development probe complementary to unique genetic sequences of target organisms
  • Automated processing, with the BD MicroProbe™ Processor (Cat. No. 23-025-600)
  • Positive results indicated by visible blue color on target organism bead
  • Built-in positive and negative controls on each results card
TRUSTED_SUSTAINABILITY
Specifications
Type MicroProbe™ Processor
For Use With (Application) Detection of vaginitis
Quantity Each
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.