Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V

Catalog No. bdb556416 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556416 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Catalog No. BDB556416 Supplier BD Supplier No. 556416
Only null left
Add to Cart
Add to Cart
Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing

For flow cytometric analysis of cells that are undergoing apoptosis

Apoptosis is a normal physiologic process which occurs during embryonic development as well as in maintenence of tissue homeostasis. The apoptotic program is characterized by certain morphologic features, including loss of plasma membrane asymmetry and attachment, condensation of cytoplasm and nucleus, and internucleosomal cleavage of DNA. In apoptotic cells, the membrane phospholipid phosphatidylserine (PS) is translocated from inner to outer leaflet of plasma membrane, thereby exposing PS to external cellular environment

  • 35-36 kDa Ca2+ dependent phospholipid-binding protein that has high affinity for PS
  • Binds to cells with exposed PS
  • May be conjugated to fluorochromes, such as fluorescein isothiocyanate (FITC), to serve as sensitive probe
  • Typically used in conjunction with vital dye such as 7-Amino-Actinomycin
  • Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide
  • Format: Purified

Flow cytometry, Blocking

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) Blocking Agent, Flow Cytometry
Formulation Aqueous Buffered solution containing ≤0.09% Sodium Azide
Molecular Weight (g/mol) 35 to 36
Name Annexin V
Quantity 0.1 mg
Storage Requirements -80°C
Recombinant Recombinant
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.