Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V, BUV395, Recombinant Protein

Catalog No. bdb564871
Click to view available options
:
100 Tests, Brilliant Ultraviolet 395

Specifications

Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Molecular Weight (g/mol) 31-35kDa
Storage Requirements 4°CXXXXX protect from prolonged exposure to light. Do not freeze.
Regulatory Status RUO
Conjugate BUV395
Cross Reactivity Human
Recombinant Recombinant
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.