Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V: FITC Apoptosis Detection Kit II

Catalog No. bdb556570 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556570 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556570 Supplier BD Supplier No. 556570
Only null left
Add to Cart
Add to Cart

Includes

Purified Recombinant Annexin V, Annexin V-FITC, Propidium Iodide Staining Solution, Annexin V Binding Buffer

Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes.

  • Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes.
  • Components: Purified Recombinant Annexin V, FITC Annexin V, Propidium Iodide Staining Solution, 10X Annexin V Binding Buffer
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage 4°C
No. of Reactions 100 Tests
For Use With (Application) Flow Cytometry
Conjugate FITC
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.