Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Anti-mouse Ig HRP Detection kit

Catalog No. bdb551011 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551011 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551011 Supplier BD Supplier No. 551011
Only null left
Add to Cart
Add to Cart

Includes

Biotinylated goat anti-mouse Igs, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, and DAB Chromogen

Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears

  • Detects molecules within a tissue microenvironment
  • Protein expression can be studied in relation to tissue or cell compartments
  • Also changes due to either normal or pathologic stimuli can be evaluated
  • Detection of an antibody bound to antigen can be accomplished by direct or indirect methods
  • Size: 200 tests
  • Regulatory status: RUO

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Storage Requirements 4°C
For Use With (Application) Immunohistochemistry, Immunohistochemical Staining
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.