Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Anti-rat Ig HRP Detection kit

Catalog No. bdb551013 Shop All BD Biosciences Products
Click to view available options
Quantity:
1 Kit

Catalog No. BDB551013

Supplier: BD 551013

Only null left
Add to Cart
Add to Cart

Includes

Biotinylated goat anti-rat Ig, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, and DAB Chromogen

Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears

  • Used with any purified primary antibody
  • Provides maximum reactivity
  • Contains protein stabilizers to enhance antibody reactivity
  • Blocking agents reduce non-specific staining

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Storage Requirements 4°C
For Use With (Application) Immunohistochemistry, Immunohistochemical Staining
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.