Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Apoptosis, DNA Damage and Cell Proliferation Kit

Catalog No. bdb562253 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562253 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562253 Supplier BD Supplier No. 562253
Only null left
Add to Cart
Add to Cart

Includes

Kit Part A:

  • PerCP-Cy5.5 Mouse Anti-BrdU
  • Alexa Fluor™ 647 Mouse Anti-H2AX (pS139)
  • PE Mouse Anti-Cleaved PARP (Asp214) Antibody
  • Cytofix/Cytoperm Fixation/Permeabilization Solution
  • Perm/Wash Buffer (10X)
  • Cytofix/Cytoperm Plus Permeabilization Buffer
  • DAPI
Kit Part B:
  • BrdU (10mg/mL)
  • DNase

Designed with the inclusion of fluorescent antibodies specific for incorporated BrdU, phosphorylated H2AX (γH2AX) and cleaved PARP

Multiparameter flow cytometry provides a powerful tool for resolving mechanisms by which individual cells in homogenous or mixed cell populations maintain viability, enter and progress through cell cycle or undergo cell death. These probes along with optimized protocols enable multicolor flow cytometric analysis of proliferation, DNA damage and apoptosis, respectively, by individual cells within samples.

  • Consists of Apoptosis, DNA Damage and Cell Proliferation Kit - Part A and Apoptosis, DNA Damage and Cell Proliferation Kit - Part B

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit Part A: 4°C and Kit Part B: -80°C
For Use With (Application) Flow Cytometry, Cell Proliferation
Quantity 50 Tests
Reactivity Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.