Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD™ Bactec™ Media: Peds Plus

Catalog No. 23028047 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
40 mL
This item is not returnable. View return policy

Catalog No. 23-028047

Supplier: BD™ 442194

This item is not returnable. View return policy

For use with BACTEC 9240, 9120, and 9050 Systems

  • Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
  • Fluorescent series bar-coded blood culture media
  • Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
  • Vials ready to use as received—require no reconstitution or dilution
  • Detection is limited to microorganisms that will grow in a particular type of medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 25°C
Form Liquid
Media Type PEDS Plus F
Product Type Microbiology Media
Formulation Processed Water 40mL, Soybean Casein digest broth 2.75% w/v, Yeast extract 0.25% w/v, Animal Tissue digest 0.10% w/v, Sodium Pyruvate 0.10% w/v, Dextrose 0.06% w/v, Sucrose 0.08% w/v, Hemin 0.0005% w/v, Menadione 0.00005% w/v, Sodium Polyanetholsulfonate (SPS) 0.020% w/v, Pyridoxal HCl (Vitamin B6) 0.001% w/v, Nonionic Adsorbing Resin 10.0% w/v, Cationic Exchange Resin 0.6% w/v
For Use With (Equipment) Bactec™ 9240, 9120 and 9050 System
No. per Pack 50
Packaging Type Vial
Quantity 40 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.