Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ BBE/CDC Anaerobe Laked Sheep Blood Agar with KV Agar Prepared Plated Media

Catalog No. l97022 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Form Liquid
Manufacturing Origin United States
Media Type BBE and CDC anaerobe Laked Sheep Blood Agar with KV
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Oxgall 15g/L, Hemin 0.01g/L, Gentamicin 0.1g/L, Vitamin K1 0.01g/L, Agar 14g/L, Growth Factors 1.8g/L, Pancreatic
For Use With (Application) Anaerobic microorganisms from clinical and nonclinical specimen
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 1220 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.