Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Chocolate II (GC II Agar) w/Hemoglobin and IsoVitaleX™/Modified Thayer Martin Agar

Catalog No. L21623 Shop All BD Diagnostic Systems Products
Click to view available options
:
Chocolate II Agar (GC II Agar) w/Hemoglobin and IsoVitaleX/Modified Thayer Martin (MTM II) Agar; 20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Chocolate II Agar

  • For the isolation and cultivation of fastidious microorganisms, Neisseria and Haemophilus species
Modified Thayer-Martin (MTM II)
  • For isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Brown
Form Liquid
Manufacturing Origin United States
Media Type Chocolate II Agar (GC II Agar) with Hemoglobin and IsoVitaleX/Modified Thayer Martin (MTM II)
Formulation Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 12g/L, Agar 10g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL, V-C-N Inhibit
For Use With (Application) Fastidious microorganisms, especially Neisseria and Haemophilus species
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 810 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.