Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Chopped Meat Carbohydrate Broth, Pre-Reduced II (Hungate Cap)
Shop All BD Diagnostic Systems Products
Click to view available options
:
Chopped Meat Carbohydrate Broth, PR II; 10/Pk.
Description
Requires
Ancillary culture media, reagents, quality control organisms and laboratory equipment
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, CMPH2 |
| Color | Beige |
| Content And Storage | 2 to 25°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Formulation | Chopped meat Pellets 10.2g/L, Pancreatic digest of Casein 30g/L, Yeast extract 5g/L, Glucose 4g/L, Dipotassium Phosphate 5g/L, Cellobiose 1g/L, Maltose 1g/L, Starch 1g/L, L-Cysteine HCl 0.5g/L, Resazurin 0.001g/L, Vitamin K1 1mg/L, Hemin 5mg/L |
| For Use With (Application) | Enrichment and cultivation of anaerobic microorganisms, obligate anaerobes |
| Packaging Type | Tube |
| Quantity | 10/Pk. |
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction