Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Columbia CNA Agar with 5% Sheep Blood/MacConkey II, I Plate™ Prepared Media

Catalog No. p-4770168
Click to view available options
Quantity:
100/Pk.
For Use With (Application):
Gram positive microorganisms
This item is not returnable. View return policy
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Columbia CNA Agar with 5% Sheep Blood

  • Selective and differential medium used for isolation and differentiation of gram-positive microorganisms
Chocolate II Agar
  • Selective and differential medium for detection of coliform organisms and enteric pathogens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Form Liquid
Manufacturing Origin United States
Media Type Columbia CNA Agar with 5% Sheep Blood/MacConkey II Agar
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Gram positive microorganisms
Packaging Type Biplate
Quantity 100/Pk.
Weight (Metric) 4350 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.