Learn More
BD BBL™ Crystal™ ID Systems
Shop All BD Diagnostic Systems ProductsDescription
Includes
Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations
Intended for identification of clinically significant aerobic and anaeorobic gram-negative and gram-positive bacterial pathogens isolated from human specimens. During incubation, the stackable 10-device trays save space and prevent incubator mess. Following incubation, distinct chromogenic and fluorogenic reactions generate a profile number, which is keyed directly into the BBL Crystal MIND Software installed on a laboratory computer.
- Onscreen display identifies the organism, reports on epidemiology, and conveys other pertinent information
- Easy-to-use systems require no pipetting or reagent addition
- Culture colonies are suspended in Crystal Broth; then inoculum suspension is poured directly into the Crystal device
- Wells fill automatically by rocking base
- Lid is snapped onto the device for safety and handling ease
- Enteric/Non-Fermenter (E/NF) Kit performs overnight identification of clinically significant aerobic gram-negative Enterobacteriaceae isolates, as well as frequently isolated glucose nonfermenting gram-negative bacilli from human origin
- Anaerobe ID (ANR) Kit is a four-hour test that identifies clinically significant anaerobes. Separate databases for Schaedler Blood Agar, CDC Anaerobe Blood Agars are included for improved specificity and increased flexibility
- Neisseria/Haemophilus (N/H) ID Kit identifies Neisseria, Haemophilus, Moraxella, Gardnerella vaginalis and other fastidious bacteria in a four-hour test
- Gram-Positive (GP) Kit identifies 121 gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in an overnight test
- Rapid Gram-Positive (RGP) Kit identifies gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in a four-hour test
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Product Type | Rapid Gram-positive ID Kit |
| For Use With (Application) | Identifies gram positive bacteria isolated from clinical specimens |
| Includes | Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations |
| Quantity | 20 Pack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.