Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase Concentrate

Catalog No. p-4897962
Click to view available options
Quantity:
20 mL
100 mL
Packaging Type:
Bottle
Tube
This item is not returnable. View return policy
This item is not returnable. View return policy

For cultivation of heterotrophic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 10000000U/mL
Additive Penicillinase
Form Liquid
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging Type Tube
Quantity 20 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.