Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ GN (Gram-Negative) Broth, 8 mL

Catalog No. bb21730 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
8 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Used for selective enrichment of Salmonella and Shigella

Inoculate broth as soon as possible after specimens arrive at laboratory.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CCAM, CMPH2, COMPF, MCM9
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type GN Broth
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
For Use With (Application) Selective enrichment of Salmonella and Shigella
No. per Pack 100
Packaging Type Tube
Quantity 8 mL
Show More Show Less

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.