Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Inhibitory Mold Agar Slants with Chloramphenicol
Shop All BD Diagnostic Systems Products
Click to view available options
:
Inhibitory Mold Agar Slant w/Chloramphenicol; 100/Pk.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Inhibitory Mold Agar |
| Formulation | Pancreatic digest of Casein 3g/L, Peptic digest of Animal Tissue 2g/L, Yeast extract 5g/L, Dextrose 5g/L, Starch 2g/L, Dextrin 1g/L, Sodium Phosphate 2g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L, Sodium Chloride 0.04g/L, Manganese Sulfate 0.16g/L, Agar 15g/L, Chloramphenicol 0.125g/L |
| For Use With (Application) | Isolation of pathogenic fungi including enhanced isolation of C. neoformans |
| Packaging Type | Tube |
| Quantity | 100 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction