Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants

Catalog No. p-4639583
Click to view available options
Quantity:
100 Pack
Packaging Type:
Tube
This item is not returnable. View return policy
This item is not returnable. View return policy

Prepared media. BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants are used in qualitative procedures for the isolation and cultivation of mycobacteria.

  • Contains a variety of inorganic salts that provide substances essential for the growth of mycobacteria
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, EP, MCM9
Color Gray
Content And Storage 2°C to 8°C
Form Highly Viscous
Manufacturing Origin United States
Media Type Middlebrook and Cohn 7H10 Agar
Formulation Magnesium Sulfate 0.05g/L, Ferric Ammonium Citrate 0.04g/L, Sodium Citrate 0.4g/L, Ammonium Sulfate 0.5g/L, Monosodium Glutamate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Agar 13.5g/L, Sodium Chloride 0.85g/L, Dextrose 2g/L, Bovine Albumin (Fraction V) 5g/L, Catalase 3mg/L, Pyridoxine 1mg/L, Zinc Sulfate 1mg/L, Copper Sulfate 1mg/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L, Malachite green 0.25mg/L, Oleic Acid 0.06mLg/L, Glycerol 5mLg/L
For Use With (Application) Qualitative procedures for the isolation and cultivation of mycobacteria
Packaging Type Tube
Quantity 100 Pack

For in vitro Diagnostic Use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.