Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ OF (Oxidation Fermentation) Media

Catalog No. p-3239950 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
Media Type:
OF media with Dextrose
This item is not returnable. View return policy
This item is not returnable. View return policy

Determines oxidative and fermentative metabolism of carbohydrates by Gram-negative rods on the basis of acid reaction

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type OF media with Dextrose
Formulation Pancreatic digest of Casein 2g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 0.3g/L, Agar 2.5g/L, Bromthymol blue 0.03g/L
For Use With (Application) Determination of oxidative and fermentative metabolism of carbohydrates by gram negative rods on the basis of Acid reaction
Packaging Type Tube
Quantity 10/Pk.

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.