Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Agar Slant

Catalog No. bb21610
Click to view available options
:
Brain heart infusion agar; 10/Pk.

For the cultivation of a wide variety of organism types, including bacteria, yeasts and molds

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Brain heart infusion agar
Formulation Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
For Use With (Application) Isolation and identification of microorganisms requiring prolonged incubation
Packaging Type Tube
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.